Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0) (16,7% Pd)
Lieferant: Thermo Scientific
|
Achtung
|
46000.MD
46000.03
46000.MDEA
114
EUR
46000.MD
46000.03
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0) (16,7% Pd)
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0)
CAS No.: 1233717-68-4
Molecular Formula: C32H56N2P2Pd
Formula Weight: 637.18
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Off-white
MDL No.: MFCD15071402
Formel:
C₃₂H₅₆N₂P₂Pd Molecular Weight: 637,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15071402 CAS-Nummer: 1233717-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...