Polyethylene glycol methyl ether acrylate, M.W. 5,000
Lieferant: Thermo Fisher Scientific
Synonyme:
mPEG-acrylate, Polyethylene glycol monomethyl ether acrylate
46388.03
46388.06
46388.03EA
102
EUR
46388.03
46388.06
Polyethylene glycol methyl ether acrylate, M.W. 5,000
Polyethylene glycol methyl ether acrylate
CAS No.: 32171-39-4
Molecular Formula: CH3(OCH2CH2)nOCOC=CH2
Storage Temperature: 2°C to 8°C
Physical Form: Granular solid
Appearance: White to off-white
MDL No.: MFCD00803685
Formel:
CH₃(OCH₂CH₂)nOCOC=CH₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD00803685 CAS-Nummer: 32171-39-4 |