Indium(III)iodid, ultra trocken ≥99,999% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr


Unter Argon in Ampullen verpackt

47108.04 47108.09 47108.18
47108.04EA 68.5 EUR
47108.04 47108.09 47108.18
Indium(III)iodid, ultra trocken ≥99,999% (Metall-Basis)
Indium(III)iodid

Purity: 99.999% CAS No.: 13510-35-5 Molecular Formula: InI3 Formula Weight: 495.53 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 4.69 Sensitivity: Light Sensitive; Hygroscopic Appearance: Yellow to brown Melting Point: 210° UN No.: UN3260 MDL No.: MFCD00016151

Formel: I₃In
Molecular Weight: 495,53 g/mol
Schmelzpunkt: 210 °C
Dichte: 4,69
Lagertemperatur: Raumtemperatur
MDL: MFCD00016151
CAS-Nummer: 13510-35-5
EINECS: 236-839-6
UN: 3260
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR