Copper bis(trifluoromethylsulphonyl)imide
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Copper bis(trifluoromethanesulphonimide), Copper bis(trifluoromethanesulphonyl)imide
47162.MD
47162.03
47162.06
47162.MDEA
48.4
EUR
47162.MD
47162.03
47162.06
Copper bis(trifluoromethylsulphonyl)imide
Copper bis(trifluoromethylsulphonyl)imide
CAS No.: 291300-50-0
Molecular Formula: C2CuF6NO4S2
Formula Weight: 343.68
Physical Form: Solid
Sensitivity: Hygroscopic
Appearance: Blue
UN No.: UN3261
MDL No.: MFCD23380170
Formel:
C₂CuF₆NO₄S₂ Molecular Weight: 343,68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23380170 CAS-Nummer: 291300-50-0 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...