Methanol 99,8 LC-MS
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Hydroxymethan, Methylalkohol, Holzalkohol
47192.K2 47192.M1
47192.K2EA 65.2 EUR
47192.K2 47192.M1
Methanol 99,8 LC-MS
Methanol
Purity: 99.8+% CAS No.: 67-56-1 Molecular Formula: CH4O Formula Weight: 32.04 Flash Point: 11°(52°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3290 Appearance: Colourless Odour: Mild Melting Point: -98° Boiling Point: 64.7° Beilstein Reference: 1098229 UN No.: UN1230 MDL No.: MFCD00004595
Applications: Solvent for LC-MS analysis
Formel: CH₃OH
Molecular Weight: 32.04 g/mol
Siedepunkt: 64.7 °C
Schmelzpunkt: –98 °C
Dichte: 0.791
Flammpunkt: 11 °C (52 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004595
CAS-Nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
Merck Index: 14,5957
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM