Zinc tetrafluoroborate hydrate (≥18% Zn)
Lieferant: Thermo Scientific
|
Gefahr
|
85124.18
85124.30
85124.09
85124.18EA
77.9
EUR
85124.18
85124.30
85124.09
Zinc tetrafluoroborate hydrate (≥18% Zn)
Zinc tetrafluoroborate hydrate
CAS No.: 27860-83-9
Molecular Formula: Zn(BF4)2.xH2O
Formula Weight: 238.98(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Odour: Odourless
UN No.: UN3260
MDL No.: MFCD00150371
Formel:
Zn(BF₄)₂·xH₂O Molecular Weight: 238,98 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150371 CAS-Nummer: 27860-83-9 EINECS: 237-534-0 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...