Iron nickel oxide, tech.
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Nickel ferrite
88530.09
88530.18
88530.09EA
64.8
EUR
88530.09
88530.18
Iron nickel oxide, tech.
Iron nickel oxide
CAS No.: 12168-54-6
Molecular Formula: NiFe2O4
Formula Weight: 234.39
Physical Form: Powder
Density: 5.368
Odour: Odourless
MDL No.: MFCD00016254
Formel:
NiFe₂O₄ Molecular Weight: 234,39 g/mol Dichte: 5,368 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016254 CAS-Nummer: 12168-54-6 EINECS: 235-335-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...