Lithium trifluoroacetate monohydrate ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
88938.09
88938.18
88938.09EA
46
EUR
88938.09
88938.18
Lithium trifluoroacetate monohydrate ≥97%
Lithium trifluoroacetate monohydrate
Purity: 97%
CAS No.: 2923-17-3
Molecular Formula: C2F3LiO2.H2O
Formula Weight: 137.97 (119.96anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
UN No.: UN2811
MDL No.: MFCD00149663
Formel:
C₂F₃LiO₂·H₂O Molecular Weight: 137,97 g/mol (119,96 anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149663 CAS-Nummer: 2923-17-3 EINECS: 220-878-0 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...