Tetraammineplatinum(II) nitrate
Lieferant: Thermo Scientific
|
Gefahr
|
88960.03
88960.06
88960.03EA
176
EUR
88960.03
88960.06
Tetraammineplatinum(II) nitrate
Tetraammineplatinum(II) nitrate
CAS No.: 20634-12-2
Molecular Formula: H12N6O6Pt
Formula Weight: 387.21
Physical Form: Powder/chunks
Appearance: White
Melting Point: 262° dec.
UN No.: UN1477
MDL No.: MFCD00011624
Formel:
H₁₂N₆O₆Pt Molecular Weight: 387,216 g/mol Schmelzpunkt: 262 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011624 CAS-Nummer: 20634-12-2 EINECS: 243-929-9 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...