(S)-Methyl-2-aminopropanoate hydrochloride 99%

Lieferant: Thermo Fisher Scientific

Synonyme: H-Ala-OMe·HCl, H-Ala-OMe.HCl, L-Alanine methyl ester hydrochloride, (S)-2-Aminopropionic Acid Methyl Ester Hydrochloride

A10178.06 A10178.14 A10178.22
A10178.06EA 36.4 EUR
A10178.06 A10178.14 A10178.22
(S)-Methyl-2-aminopropanoate hydrochloride 99%
(S)-Methyl-2-aminopropanoate hydrochloride

Purity: 99% CAS No.: 2491-20-5 Molecular Formula: C4H9NO2.HCl Formula Weight: 139.58 Storage Temperature: 2°C to 8°C Melting Point: 106-110° Beilstein Reference: 3594033 Optical Rotation: +6.5° (c=10 in methanol) MDL No.: MFCD00063663

Formel: C₄H₉NO₂·HCl
Molecular Weight: 139.6 g/mol
Schmelzpunkt: 106…110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00063663
CAS-Nummer: 2491-20-5
EINECS: 219-652-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR