3-Aminophthalic acid hydrochloride dihydrate 98%

Lieferant: Thermo Scientific
Achtung

A10319.03 A10319.06 A10319.14
A10319.03EA 24.7 EUR
A10319.03 A10319.06 A10319.14
3-Aminophthalic acid hydrochloride dihydrate 98%
3-Aminophthalic acid hydrochloride dihydrate

Purity: 98% CAS No.: 6946-22-1 Molecular Formula: C8H7NO4.HCl.2H2O Formula Weight: 253.65 (217.62anhy) Melting Point: 182° dec. MDL No.: MFCD00150282

Formel: C₈H₇NO₄·HCl·2H₂O
Molecular Weight: 253,65 g/mol (217,62 anhy)
Schmelzpunkt: 182 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150282
CAS-Nummer: 6946-22-1
EINECS: 230-106-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR