(2E)-3-[4-(Acetyloxy)-3-methoxyphenyl]acrylic acid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A10361.03
A10361.06
A10361.14
A10361.03EA
32.4
EUR
A10361.03
A10361.06
A10361.14
(2E)-3-[4-(Acetyloxy)-3-methoxyphenyl]acrylic acid ≥98%
(2E)-3-[4-(Acetyloxy)-3-methoxyphenyl]acrylic acid
Purity: 98+%
CAS No.: 2596-47-6
Molecular Formula: C12H12O5
Formula Weight: 236.22
Odour: Odourless
Melting Point: 193-196°
Beilstein Reference: 2218026
MDL No.: MFCD00014378
Formel:
C₁₂H₁₂O₅ Schmelzpunkt: 193…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014378 CAS-Nummer: 2596-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...