Cholesteryl chloride 98%

Lieferant: Thermo Scientific

Synonyme: Cholesteryl Chloride, (3S,8S,9S,10R,13R,14S,17R)-3-Chloro-10,13-dimethyl-17-((R)-6-methylheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene

A10433.14 A10433.22
A10433.14EA 60 EUR
A10433.14 A10433.22
Cholesteryl chloride 98%
Cholesteryl chloride

Purity: 98% CAS No.: 910-31-6 Molecular Formula: C27H45Cl Formula Weight: 405.11 Melting Point: 94-96° Beilstein Reference: 2703655 Optical Rotation: -26° (c=1 in benzene) MDL No.: MFCD00003632

Formel: C₂₇H₄₅Cl
Molecular Weight: 405,107 g/mol
Schmelzpunkt: 94…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003632
CAS-Nummer: 910-31-6
EINECS: 213-004-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR