Diethylammonium-diethyldithiocarbamat 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Diethyldithiocarbamic Acid Diethylammonium Salt, Diethylammonium diethylcarbamodithioate

A10458.14 A10458.06 A10458.22
A10458.14EA 125 EUR
A10458.14 A10458.06 A10458.22
Diethylammonium-diethyldithiocarbamat 98%
Diethylammonium diethyldithiocarbamate

Purity: 98% CAS No.: 1518-58-7 Molecular Formula: C9H22N2S2 Formula Weight: 222.42 Melting Point: 81-84° MDL No.: MFCD00013146

Formel: C₅H₁₁NS₂
Molecular Weight: 222,41 g/mol
Schmelzpunkt: 81…84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013146
CAS-Nummer: 1518-58-7
EINECS: 216-175-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR