7,7,8,8-Tetracyanoquinodimethane 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1,4-Bis(dicyanomethylene)cyclohexadiene, TCNQ, 7,7,8,8-Tetracyanoquinodimethane, 7,7,8,8-Tetracyanoquinonedimethane

A10779.06 A10779.14
A10779.06EA 119 EUR
A10779.06 A10779.14
7,7,8,8-Tetracyanoquinodimethane 98%
7,7,8,8-Tetracyanoquinodimethane

Purity: 98% CAS No.: 1518-16-7 Molecular Formula: C12H4N4 Formula Weight: 204.19 Melting Point: ca 288° dec. Beilstein Reference: 611604 UN No.: UN3439 MDL No.: MFCD00011664

Formel: C₁₂H₄N₄
Molecular Weight: 204.19 g/mol
Schmelzpunkt: ∼ 288 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011664
CAS-Nummer: 1518-16-7
EINECS: 216-174-8
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR