N-α-Benzoyl-L-arginine 99%

Lieferant: Thermo Fisher Scientific

Synonyme: Bz-Arg-OH, N-α-Benzoyl-L-Arginine

A10812.09 A10812.18
A10812.09EA 29.9 EUR
A10812.09 A10812.18
N-α-Benzoyl-L-arginine 99%
N-alpha-Benzoyl-L-arginine

Purity: 99% CAS No.: 154-92-7 Molecular Formula: C13H18N4O3 Formula Weight: 278.31 Storage Temperature: 2°C to 8°C Melting Point: 285° dec. MDL No.: MFCD00001763

Formel: C₁₃H₁₈N₄O₃
Molecular Weight: 278.312 g/mol
Schmelzpunkt: 285 °C (Zers.)
MDL: MFCD00001763
CAS-Nummer: 154-92-7
EINECS: 205-837-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR