3-Amino-1-phenyl-2-pyrazolin-5-one 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
5-Amino-2,4-dihydro-2-phenyl-3H-pyrazol-3-one
A10862.14
A10862.22
A10862.14EA
27.1
EUR
A10862.14
A10862.22
3-Amino-1-phenyl-2-pyrazolin-5-one 97%
3-Amino-1-phenyl-2-pyrazolin-5-one
Purity: 97%
CAS No.: 4149-06-8
Molecular Formula: C9H9N3O
Formula Weight: 175.19
Melting Point: ca 213° dec.
Beilstein Reference: 153681
MDL No.: MFCD00003137
Formel:
C₉H₉N₃O Molecular Weight: 175.19 g/mol Schmelzpunkt: ∼ 213 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003137 CAS-Nummer: 4149-06-8 EINECS: 223-974-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...