Isonicotinoylchloridhydrochlorid 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Pyridine-4-carbonyl Chloride Hydrochloride, 4-Pyridinoyl chloride hydrochloride
A10863.09
A10863.18
A10863.30
A10863.09EA
43.2
EUR
A10863.09
A10863.18
A10863.30
Isonicotinoylchloridhydrochlorid 98%
Isonicotinoyl chloride hydrochloride
Purity: 98%
CAS No.: 39178-35-3
Molecular Formula: C6H4ClNO.HCl
Formula Weight: 178.01
Melting Point: 154-162°
Beilstein Reference: 3626052
UN No.: UN3261
MDL No.: MFCD00012830
Formel:
C₆H₄ClNO Molecular Weight: 178.01 g/mol Schmelzpunkt: 154…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012830 CAS-Nummer: 39178-35-3 EINECS: 254-331-2 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...