2,4-Dimethyl-6-hydroxypyrimidine 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,6-Dimethylpyrimidin-4-ol, 2,6-Dimethyl-4-pyrimidinol, 6-Hydroxy-2,4-dimethylpyrimidine

A10960.06 A10960.14
A10960.06EA 26.5 EUR
A10960.06 A10960.14
2,4-Dimethyl-6-hydroxypyrimidine 99%
2,4-Dimethyl-6-hydroxypyrimidine

Purity: 99% CAS No.: 6622-92-0 Molecular Formula: C6H8N2O Formula Weight: 124.14 Melting Point: 198-200° Beilstein Reference: 114324 MDL No.: MFCD00006105

Formel: C₆H₈N₂O
Molecular Weight: 124,14 g/mol
Schmelzpunkt: 198…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006105
CAS-Nummer: 6622-92-0
EINECS: 229-577-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR