Hemin chloride (aus Schwein) 97+%

Lieferant: Thermo Scientific

Synonyme: Chlorohemin, 1,3,5,8-Tetramethyl-2,4-divinylporphine-6,7-dipropionic acid ferrichloride, Chloroprotoporphyrin IX iron(III), Haemin, Hemin, Ferriprotoporphyrin IX chloride

A11165.06 A11165.03 A11165.14
A11165.06EA 140 EUR
A11165.06 A11165.03 A11165.14
Hemin chloride (aus Schwein) 97+%
Hemin chloride

Stimulates synthesis of globulin.


Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.


Soluble in dilute ammonia, trimethylamine, p-toluidine and dimethylaniline. Slightly soluble in alcohol. Insoluble in water


Bitte beachten Incompatible with strong oxidizing agents.

Formel: C₃₄H₃₂ClFeN₄O₄
Molecular Weight: 651,95 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00010726
CAS-Nummer: 16009-13-5
EINECS: 240-140-1
Merck Index: 14,4644

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR