Methyl-p-tolylsulfon 99%

Lieferant: Thermo Scientific

Synonyme: 4-(Methylsulphonyl)toluene, 1-Methyl-4-(methylsulphonyl)benzene

A11531.06 A11531.14
A11531.06EA 53.6 EUR
A11531.06 A11531.14
Methyl-p-tolylsulfon 99%
Methyl-p-tolylsulfon

Purity: 99% CAS No.: 3185-99-7 Molecular Formula: C8H10O2S Formula Weight: 170.23 Melting Point: 86-89° Boiling Point: 140°/3mm Beilstein Reference: 607296 MDL No.: MFCD00014742

Formel: C₈H₁₀O₂S
Molecular Weight: 170,23 g/mol
Siedepunkt: 140 °C (3 mmHg)
Schmelzpunkt: 86…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014742
CAS-Nummer: 3185-99-7
EINECS: 221-682-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR