Methyl-α-D-mannopyranosid ≥99%
Lieferant: Thermo Scientific
Synonyme:
ɑ-Methyl-D-mannoside, Methyl-ɑ-D-Mannoside, Methyl α-D-Mannopyranoside, ɑ-Methyl-D-mannopyranoside, (2R,3S,4S,5S,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
A11533.14
A11533.22
A11533.14EA
44.7
EUR
A11533.14
A11533.22
Methyl-α-D-mannopyranosid ≥99%
Methyl-α-D-mannopyranosid
Purity: 99%
CAS No.: 617-04-9
Molecular Formula: C7H14O6
Formula Weight: 194.18
Melting Point: 193-196°
Beilstein Reference: 81566
Optical Rotation: +79° (c=10 in water)
MDL No.: MFCD00063262
Formel:
C₇H₁₄O₆ Molecular Weight: 194,18 g/mol Schmelzpunkt: 193…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063262 CAS-Nummer: 617-04-9 EINECS: 210-502-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...