Sodium-DL-3-hydroxybutyrate 98%

Lieferant: Thermo Scientific

Synonyme: Sodium 3-hydroxybutanoate, DL-3-Hydroxybutyric acid Sodium salt

A11613.06 A11613.14
A11613.06EA 36.9 EUR
A11613.06 A11613.14
Sodium-DL-3-hydroxybutyrate 98%
Sodium-DL-3-hydroxybutyrate

Purity: 98% CAS No.: 150-83-4 Molecular Formula: C4H7NaO3 Formula Weight: 126.09 Melting Point: 161-167° Beilstein Reference: 4824866 MDL No.: MFCD00016716

Formel: C₄H₈O₃
Molecular Weight: 126,09 g/mol
Schmelzpunkt: 161…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016716
CAS-Nummer: 150-83-4
EINECS: 205-774-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR