5-Hydantoin acetic acid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11731.06
A11731.14
A11731.06EA
32.4
EUR
A11731.06
A11731.14
5-Hydantoin acetic acid 98%
5-Hydantoin acetic acid
Purity: 98%
CAS No.: 5427-26-9
Molecular Formula: C5H6N2O4
Formula Weight: 158.11
Melting Point: ca 217° dec.
Beilstein Reference: 83035
MDL No.: MFCD00005267
Formel:
C₅H₆N₂O₄ Molecular Weight: 158.113 g/mol Schmelzpunkt: ∼ 217 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005267 CAS-Nummer: 5427-26-9 EINECS: 226-574-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...