4,6-Diphenyl-2-pyrone 98%

Lieferant: Thermo Scientific

A11755.03 A11755.06
A11755.03EA 60.8 EUR
A11755.03 A11755.06
4,6-Diphenyl-2-pyrone 98%
4,6-Diphenyl-2-pyrone

Purity: 98% CAS No.: 17372-52-0 Molecular Formula: C17H12O2 Formula Weight: 248.27 Odour: Odourless Melting Point: 135-138° MDL No.: MFCD00031018

Formel: C₁₇H₁₂O₂
Molecular Weight: 248,27 g/mol
Schmelzpunkt: 135…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031018
CAS-Nummer: 17372-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR