m-Toluoylacetonitrile 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Oxo-3-(m-tolyl)propanenitrile
A11965.03
A11965.06
A11965.03EA
26.6
EUR
A11965.03
A11965.06
m-Toluoylacetonitrile 97%
m-Toluoylacetonitrile
Purity: 97%
CAS No.: 53882-81-8
Molecular Formula: C10H9NO
Formula Weight: 159.19
Melting Point: 71-75°
Beilstein Reference: 2717335
MDL No.: MFCD00067922
Formel:
C₁₀H₉NO Molecular Weight: 159,19 g/mol Schmelzpunkt: 71…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067922 CAS-Nummer: 53882-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...