L-Leucine-p-nitroanilide 99%
Lieferant: Thermo Scientific
Synonyme:
H-Leu-pNA, (S)-2-Amino-4-methyl-N-(4-nitrophenyl)pentanamide
A11994.03
A11994.06
A11994.03EA
57
EUR
A11994.03
A11994.06
L-Leucine-p-nitroanilide 99%
L-Leucine-p-nitroanilide
Purity: 99%
CAS No.: 4178-93-2
Molecular Formula: C12H17N3O3
Formula Weight: 251.29
Storage Temperature: 2°C to 8°C
Melting Point: 89-90°
Beilstein Reference: 2218433
MDL No.: MFCD00063681
Formel:
C₁₂H₁₇N₃O₃ Molecular Weight: 251,286 g/mol Schmelzpunkt: 89…90 °C |
MDL:
MFCD00063681 CAS-Nummer: 4178-93-2 EINECS: 224-047-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...