Di-2-thienyl ketone 98%

Lieferant: Thermo Scientific

Synonyme: 2,2'-Carbonyldi(thiophene), Di(thiophen-2-yl)methanone

A12062.03 A12062.06 A12062.14
A12062.03EA 61.5 EUR
A12062.03 A12062.06 A12062.14
Di-2-thienyl ketone 98%
Di-2-thienyl ketone

Purity: 98% CAS No.: 704-38-1 Molecular Formula: C9H6OS2 Formula Weight: 194.28 Melting Point: 87-90° Beilstein Reference: 129168 MDL No.: MFCD00052096

Formel: C₉H₆OS₂
Molecular Weight: 194,28 g/mol
Schmelzpunkt: 87…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052096
CAS-Nummer: 704-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR