Penta-O-acetyl-β-D-galactopyranose 98%

Lieferant: Thermo Scientific

Synonyme: β-D-Galactose Pentaacetate, (2S,3R,4S,5S,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

A12163.14 A12163.22
A12163.14EA 71.3 EUR
A12163.14 A12163.22
Penta-O-acetyl-β-D-galactopyranose 98%
Penta-O-acetyl-beta-D-galactopyranose

Purity: 98% CAS No.: 4163-60-4 Molecular Formula: C16H22O11 Formula Weight: 390.34 Melting Point: 142-146° Beilstein Reference: 98853 Optical Rotation: +24° (c=4 in chloroform) MDL No.: MFCD00063259

Formel: C₁₆H₂₂O₁₁
Molecular Weight: 390,34 g/mol
Schmelzpunkt: 142…146 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063259
CAS-Nummer: 4163-60-4
EINECS: 224-008-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR