2-Chloro-N-(hydroxymethyl)acetamide 98%
Lieferant: Thermo Scientific
|
Gefahr
|
A12247.09
A12247.18
A12247.09EA
67.5
EUR
A12247.09
A12247.18
2-Chloro-N-(hydroxymethyl)acetamide 98%
2-Chloro-N-(hydroxymethyl)acetamide
Purity: 98%
CAS No.: 2832-19-1
Molecular Formula: C3H6ClNO2
Formula Weight: 123.54
Storage Temperature: 2°C to 8°C
Melting Point: 97-103°
Beilstein Reference: 1811765
UN No.: UN3261
MDL No.: MFCD00021961
Formel:
C₃H₆ClNO₂ Molecular Weight: 123,54 g/mol Schmelzpunkt: 97…103 °C |
MDL:
MFCD00021961 CAS-Nummer: 2832-19-1 EINECS: 220-598-9 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...