1-Benzothiophene-2-carboxamide 97%

Lieferant: Thermo Fisher Scientific

A12292.03 A12292.06
A12292.03EA 40 EUR
A12292.03 A12292.06
1-Benzothiophene-2-carboxamide 97%
1-Benzothiophene-2-carboxamide

Purity: 97% CAS No.: 6314-42-7 Molecular Formula: C9H7NOS Formula Weight: 177.23 Melting Point: 176-178° Beilstein Reference: 4467 MDL No.: MFCD00052505

Formel: C₉H₇NOS
Schmelzpunkt: 176…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052505
CAS-Nummer: 6314-42-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR