Sulphadiazine 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-(2-Pyrimidinyl)sulphanilamide, 4-Amino-N-(pyrimidin-2-yl)benzenesulphonamide, 4-Amino-N-(2-pyrimidinyl)benzenesulphonamide

A12370.18 A12370.0B A12370.30
A12370.18EA 28.9 EUR
A12370.18 A12370.0B A12370.30
Sulphadiazine 99%
Sulphadiazine

Purity: 99% CAS No.: 68-35-9 Molecular Formula: C10H10N4O2S Formula Weight: 250.28 Physical Form: Powder Appearance: White Melting Point: 253-256° MDL No.: MFCD00006065

Formel: C₁₀H₁₀N₄O₂S
Molecular Weight: 250,28 g/mol
Schmelzpunkt: 253…256 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006065
CAS-Nummer: 68-35-9
EINECS: 200-685-8
Merck Index: 14,8903

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR