Phosphoenolpyruvic acid monocyclohexylammonium salt 98%
Lieferant: Thermo Scientific
Synonyme:
PEP·CHX
A12385.03
A12385.14
A12385.06
A12385.03EA
82
EUR
A12385.03
A12385.14
A12385.06
Phosphoenolpyruvic acid monocyclohexylammonium salt 98%
Phosphoenolpyruvic acid monocyclohexylammonium salt
Purity: 98%
CAS No.: 10526-80-4
Molecular Formula: C9H18NO6P
Formula Weight: 267.22
Storage Temperature: -30°C to -10°C
Melting Point: ca 140° dec.
MDL No.: MFCD00036375
Formel:
C₃H₅O₆P Molecular Weight: 267,22 g/mol Schmelzpunkt: ∼ 140 °C (Zersetzung) |
MDL:
MFCD00036375 CAS-Nummer: 10526-80-4 EINECS: 234-084-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...