Ethyl-4-pentenoat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ethyl pent-4-enoate, 4-Pentenoic Acid Ethyl Ester
A12422.03
A12422.06
A12422.14
A12422.22
A12422.03EA
29.3
EUR
A12422.03
A12422.06
A12422.14
A12422.22
Ethyl-4-pentenoat ≥98%
Ethyl-4-pentenoate
Purity: 98+%
CAS No.: 1968-40-7
Molecular Formula: CH2=CHCH2CH2CO2CH2CH3
Formula Weight: 128.17
Flash Point: 37°(98°F)
Refractive Index: 1.4170
Boiling Point: 146°
Beilstein Reference: 1751643
UN No.: UN3272
MDL No.: MFCD00036562
Formel:
C₇H₁₂O₂ Molecular Weight: 128,17 g/mol Siedepunkt: 146 °C Dichte: 0,900 Flammpunkt: 37 °C (98 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036562 CAS-Nummer: 1968-40-7 EINECS: 217-818-0 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...