3-Phenyl-1,2,4-triazole-5-thiol hydrate 98%

Lieferant: Thermo Fisher Scientific
Achtung

A12427.03 A12427.06
A12427.03EA 23.5 EUR
A12427.03 A12427.06
3-Phenyl-1,2,4-triazole-5-thiol hydrate 98%
3-Phenyl-1,2,4-triazole-5-thiol hydrate

Purity: 98% CAS No.: 3414-94-6 Molecular Formula: C8H7N3S.xH2O Formula Weight: 177.23(anhy) Melting Point: 253-258° Beilstein Reference: 135556 MDL No.: MFCD00150547

Formel: C₈H₇N₃S·xH₂O
Molecular Weight: 177,23 g/mol (anhy)
Schmelzpunkt: 253…258 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150547
CAS-Nummer: 3414-94-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR