2-Hydroxy-4,6-dimethylnicotinonitril 98%

Lieferant: Thermo Scientific
Gefahr

A12466.14 A12466.22
A12466.14EA 39.1 EUR
A12466.14 A12466.22
2-Hydroxy-4,6-dimethylnicotinonitril 98%
4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

Purity: 98% CAS No.: 769-28-8 Molecular Formula: C8H8N2O Formula Weight: 148.17 Melting Point: ca 290° dec. Beilstein Reference: 130992 MDL No.: MFCD00006269

Formel: C₈H₈N₂O
Molecular Weight: 148,17 g/mol
Schmelzpunkt: ∼ 290 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006269
CAS-Nummer: 769-28-8
EINECS: 212-207-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR