Penta-O-acetyl-β-D-glucopyranose 98%
Lieferant: Thermo Scientific
Synonyme:
β-D-Glucose Pentaacetate, Pentaacetyl-β-D-glucose, B-D-Glucose Pentaacetate, (2S,3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate
A12526.0B
A12526.18
A12526.30
A12526.0BEA
397
EUR
A12526.0B
A12526.18
A12526.30
Penta-O-acetyl-β-D-glucopyranose 98%
Penta-O-acetyl-beta-D-glucopyranose
Purity: 98%
CAS No.: 604-69-3
Molecular Formula: C16H22O11
Formula Weight: 390.34
Melting Point: 130-132°
Beilstein Reference: 98851
Optical Rotation: +5° (c=5 in chloroform)
MDL No.: MFCD00006597
Formel:
C₁₆H₂₂O₁₁ Molecular Weight: 390,34 g/mol Schmelzpunkt: 130…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006597 CAS-Nummer: 604-69-3 EINECS: 210-074-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...