3-Ethoxycarbonylcoumarin 98%
Lieferant: Thermo Scientific
Synonyme:
Ethyl Coumarin-3-carboxylate, Coumarin-3-carboxylic acid ethyl ester
A12553.03
A12553.06
A12553.03EA
25.3
EUR
A12553.03
A12553.06
3-Ethoxycarbonylcoumarin 98%
3-Ethoxycarbonylcoumarin
Purity: 98%
CAS No.: 1846-76-0
Molecular Formula: C12H10O4
Formula Weight: 218.21
Melting Point: 92-95°
Beilstein Reference: 200147
MDL No.: MFCD00016964
Formel:
C₁₂H₁₀O₄ Molecular Weight: 218,21 g/mol Schmelzpunkt: 92…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016964 CAS-Nummer: 1846-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...