Chinidin

Lieferant: Thermo Scientific
Gefahr

Synonyme: (+)-Quinidine

A12559.03 A12559.06 A12559.14
A12559.03EA 25.7 EUR
A12559.03 A12559.06 A12559.14
Chinidin
Quinidine

CAS No.: 56-54-2 Molecular Formula: C20H24N2O2 Formula Weight: 324.42 Melting Point: 171-173° Beilstein Reference: 91866 Optical Rotation: +260° (c=1 in ethanol) UN No.: UN2811 MDL No.: MFCD00135581


Applications: Antiarrhythmic agent that blocks the inward sodium channel

Formel: C20H24N2O2
Molecular Weight: 324,42 g/mol
Schmelzpunkt: 171…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00135581
CAS-Nummer: 56-54-2
EINECS: 200-279-0
ADR: 6.1,III
Merck Index: 14,8060

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR