3-(Phenylthio)thiophene 97%

Lieferant: Thermo Scientific
Gefahr

A12585.03 A12585.06
A12585.03EA 65.6 EUR
A12585.03 A12585.06
3-(Phenylthio)thiophene 97%
3-(Phenylthio)thiophene

Purity: 97% CAS No.: 16718-11-9 Molecular Formula: C10H8S2 Formula Weight: 192.30 Boiling Point: 111-112°/0.6mm MDL No.: MFCD00052043

Formel: C₁₀H₈S₂
Molecular Weight: 192,30 g/mol
Siedepunkt: 111…112 °C (0,6 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052043
CAS-Nummer: 16718-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR