Methyl-2-thiophencarboxylat 97%

Lieferant: Thermo Scientific

Synonyme: Methyl thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Methyl Ester

A12677.14 A12677.06 A12677.22
A12677.14EA 183 EUR
A12677.14 A12677.06 A12677.22
Methyl-2-thiophencarboxylat 97%
Methyl-2-thiophencarboxylat

Purity: 97% CAS No.: 5380-42-7 Molecular Formula: C6H6O2S Formula Weight: 142.18 Refractive Index: 1.5420 Boiling Point: 95-97°/15mm Beilstein Reference: 111111 MDL No.: MFCD00016895

Formel: C₆H₆O₂S
Molecular Weight: 142,17 g/mol
Siedepunkt: 95…97 °C (15 mmHg)
Dichte: 1,230
Lagertemperatur: Raumtemperatur
MDL: MFCD00016895
CAS-Nummer: 5380-42-7
EINECS: 226-371-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR