Ethyl-(triphenylphosphoranyliden)acetat ≥98%
Lieferant: Thermo Scientific
Synonyme:
Ethyl triphenylphosphoranylidene-acetate, (Triphenylphosphoranylidene)acetic acid ethyl ester, (Carbethoxymethylene)triphenylphosphorane, Ethoxycarbonylmethylene-triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)acetate
A12896.09
A12896.18
A12896.30
A12896.09EA
39.5
EUR
A12896.09
A12896.18
A12896.30
Ethyl-(triphenylphosphoranyliden)acetat ≥98%
Ethyl-2-(triphenylphosphoranylidene)acetate
Purity: 98+%
CAS No.: 1099-45-2
Molecular Formula: CH3CH2O2CCH=P(C6H5)3
Formula Weight: 348.38
Storage Temperature: 2°C to 8°C
Melting Point: 126-130°
Beilstein Reference: 754639
MDL No.: MFCD00009183
Formel:
C₂₂H₂₁O₂P Molecular Weight: 348,38 g/mol Schmelzpunkt: 126…130 °C |
MDL:
MFCD00009183 CAS-Nummer: 1099-45-2 EINECS: 214-151-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...