3-(2-Furyl)acrolein 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-(Furan-2-yl)acrylaldehyde, 3-(2-Furyl)propenal

A13042.06 A13042.14
A13042.06EA 26.1 EUR
A13042.06 A13042.14
3-(2-Furyl)acrolein 99%
3-(2-Furyl)acrolein

Purity: 99% CAS No.: 623-30-3 Molecular Formula: C7H6O2 Formula Weight: 122.12 Storage Temperature: 2°C to 8°C Flash Point: 99°(210°F) Melting Point: 49-52° Boiling Point: 95°/5mm Beilstein Reference: 107570 UN No.: UN1759 MDL No.: MFCD00003256

Formel: C₇H₆O₂
Molecular Weight: 122,12 g/mol
Siedepunkt: 95 °C (5 mmHg)
Schmelzpunkt: 49…52 °C
Flammpunkt: 99 °C (210 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00003256
CAS-Nummer: 623-30-3
EINECS: 210-785-3
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR