Methyl-2-(aminosulfonyl)benzoat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 2-sulphamoylbenzoate, 2-sulphamoylbenzoic acid Methyl Ester, Methyl 2-(Aminosulphonyl)benzoate, 2-(Aminosulphonyl)benzoic acid Methyl Ester, 2-sulphonamidobenzoic acid Methyl Ester, Methyl 2-sulphonamidobenzoate

A13553.22 A13553.14
A13553.22EA 105 EUR
A13553.22 A13553.14
Methyl-2-(aminosulfonyl)benzoat 98%
Methyl-2-(aminosulphonyl)benzoate

Purity: 98% CAS No.: 57683-71-3 Molecular Formula: C8H9NO4S Formula Weight: 215.23 Melting Point: 123-125° MDL No.: MFCD00009808

Formel: C₈H₉NO₄S
Molecular Weight: 215.22 g/mol
Schmelzpunkt: 123…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009808
CAS-Nummer: 57683-71-3
EINECS: 260-903-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR