N-[(Benzyloxy)carbonyl]-L-valin 99%
Lieferant: Thermo Scientific
Synonyme:
N-Cbz-L-valine, Z-L-Val-OH, Z-Val-OH
A13583.14
A13583.22
A13583.14EA
106
EUR
A13583.14
A13583.22
N-[(Benzyloxy)carbonyl]-L-valin 99%
N-Carbobenzyloxy-L-valine
Purity: 99%
CAS No.: 1149-26-4
Molecular Formula: C13H17NO4
Formula Weight: 251.28
Melting Point: 60°
Beilstein Reference: 2056617
MDL No.: MFCD00008922
Formel:
C₁₃H₁₇NO₄ Molecular Weight: 251,3 g/mol Schmelzpunkt: 60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008922 CAS-Nummer: 1149-26-4 EINECS: 214-562-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...