4-Methoxycoumarin 98%

Lieferant: Thermo Scientific

A13670.06 A13670.14
A13670.06EA 60.4 EUR
A13670.06 A13670.14
4-Methoxycoumarin 98%
4-Methoxycoumarin

Purity: 98% CAS No.: 20280-81-3 Molecular Formula: C10H8O3 Formula Weight: 176.17 Melting Point: 122-125° Beilstein Reference: 145960 MDL No.: MFCD00017349

Formel: C₁₀H₈O₃
Molecular Weight: 176,17 g/mol
Schmelzpunkt: 122…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017349
CAS-Nummer: 20280-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR