Diethyl-bis(hydroxymethyl)malonate 97%

Lieferant: Thermo Fisher Scientific

Synonyme: Diethyl 2,2-bis(hydroxymethyl)malonate

A13677.14 A13677.22
A13677.14EA 52.7 EUR
A13677.14 A13677.22
Diethyl-bis(hydroxymethyl)malonate 97%
Diethyl-bis(hydroxymethyl)malonate

Purity: 97% CAS No.: 20605-01-0 Molecular Formula: C9H16O6 Formula Weight: 220.22 Flash Point: 110°(230°F) Melting Point: 47-52° Beilstein Reference: 1788515 MDL No.: MFCD00009130

Formel: C₉H₁₆O₆
Molecular Weight: 220.221 g/mol
Schmelzpunkt: 47…52 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009130
CAS-Nummer: 20605-01-0
EINECS: 243-916-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR