4-Piperidone monohydrate hydrochloride 98%
Lieferant: Thermo Scientific
Synonyme: 4,4-Piperidinediol Hydrochloride, 4,4-Dihydroxypiperidine Hydrochloride, 4-Ketopiperidine Monohydrate Hydrochloride, 4-Piperidinone monohydrate hydrochloride, Piperidin-4-one hydrate hydrochloride, 4-Azacyclohexanone Monohydrate Hydrochloride
A13726.09 A13726.18
A13726.09EA 39.6 EUR
A13726.09 A13726.18
4-Piperidone monohydrate hydrochloride 98%
4-Piperidone monohydrate hydrochloride
Purity: 98% CAS No.: 40064-34-4 Molecular Formula: C5H9NO.HCl.H2O Formula Weight: 153.61 (135.59anhy) Melting Point: 94-100° Beilstein Reference: 3916558 MDL No.: MFCD00012776
Formel: C₅H₉NO
Molecular Weight: 99,13 g/mol (free base)
Schmelzpunkt: 94…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012776
CAS-Nummer: 40064-34-4
EINECS: 254-779-9