4-Hydroxy-3,5-dimethoxybenzyl alcohol 97%

Lieferant: Thermo Scientific
Achtung

A13888.03 A13888.06 A13888.14
A13888.03EA 25.4 EUR
A13888.03 A13888.06 A13888.14
4-Hydroxy-3,5-dimethoxybenzyl alcohol 97%
4-Hydroxy-3,5-dimethoxybenzyl alcohol

Purity: 97% CAS No.: 530-56-3 Molecular Formula: C9H12O4 Formula Weight: 184.19 Melting Point: 129-132° Beilstein Reference: 1873975 MDL No.: MFCD00016871

Formel: C₉H₁₂O₄
Molecular Weight: 184,19 g/mol
Schmelzpunkt: 129…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016871
CAS-Nummer: 530-56-3
EINECS: 208-485-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR