Methyl-(triphenylphosphoranyliden)acetat 98%
Lieferant: Thermo Scientific
Synonyme:
Methyl (triphenylphosphoranylidene)acetate, Methyl 2-(triphenylphosphoranylidene)acetate, Methoxycarbonylmethylene-Triphenylphosphorane
A14020.14
A14020.22
A14020.36
A14020.14EA
101
EUR
A14020.14
A14020.22
A14020.36
Methyl-(triphenylphosphoranyliden)acetat 98%
Methyl-2-(triphenylphosphoranylidene)acetate
Purity: 98%
CAS No.: 2605-67-6
Molecular Formula: CH3O2CCH=P(C6H5)3
Formula Weight: 334.36
Melting Point: 165-169°
Beilstein Reference: 618430
MDL No.: MFCD00008455
Formel:
C₂₁H₁₉O₂P Molecular Weight: 334,36 g/mol Schmelzpunkt: 165…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008455 CAS-Nummer: 2605-67-6 EINECS: 220-018-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...